missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphoranylidene)ammonium Chloride, TRC

Product Code. 30332084
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332084 Supplier Toronto Research Chemicals Supplier No. TRCB5887105G
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(triphenylphosphoranylidene)ammonium Chloride
CAS 21050-13-5
Molecular Formula C36H30ClNP2
Quantity 5 g
Synonym Triphenyl(P,P,P-triphenylphosphine imidato)-phosphorus Chloride, (T-4)-Triphenyl(P,P,P-triphenylphosphine imidato-N)-phosphorus Chloride, P,P,P-triphenyl-Phosphine Imide Phosphorus Complex
InChI Formula InChI=1S/C36H30NP2.ClH/c1-7-19-31(20-8-1)38(32-21-9-2-10-22-32,33-23-11-3-12-24-33)37-39(34-25-13-4-14-26-34,35-27-15-5-16-28-35)36-29-17-6-18-30-36;/h1-30H;1H/q+1;/p-1
SMILES C1(P(C2=CC=CC=C2)(C3=CC=CC=C3)=[N+]=P(C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6)=CC=CC=C1.[Cl-]
IUPAC Name 1,1,1-triphenyl-N-(triphenyl-l5-phosphaneylidene)-l5-phosphaniminium chloride
Molecular Weight (g/mol) 574.03
Formula Weight 573.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.