missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)iminium trifluoroacetate, 98%, Thermo Scientific™

Product Code. 15416596
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Bis(triphenylphosphine)iminium trifluoroacetate is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116405-43-7
Molecular Formula C38H30F3NO2P2
Molecular Weight (g/mol) 651.61
MDL Number MFCD15146355
InChI Key NDLHFNFVQBXEPE-UHFFFAOYSA-M
Synonym 1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium 2,2,2-trifluoroacetate, ppn tfa, ksc924o3n, bis triphenylphosphine iminiumtrifluoroacetate, bis-triphenylphosphoranylidene ammonium tfa, bis triphenylphosphine iminium trifluoroacetic acid salt, bis-triphenylphosphoranylidene ammonium tfa ppn tfa, bis triphenyl-??-phosphanylidene azanium trifluoroacetate, 1,1,1-triphenyl-n-triphenylphosphoranylidene-phosphoraniminium 2,2,2-trifluoroacetate, triphenyl-n-triphenyl-lambda∼5∼-phosphanylidene-lambda∼5∼-phosphaniminium trifluoroacetate
PubChem CID 19800386
IUPAC Name 2,2,2-trifluoroacetate;triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium
SMILES [O-]C(=O)C(F)(F)F.C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 782.65
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Bis(triphenylphosphine)iminium trifluoroacetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'