missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)iminium trifluoroacetate, 98%, Thermo Scientific™

Product Code. 15426596
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15426596

Brand: Thermo Scientific Alfa Aesar H61550.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Bis(triphenylphosphine)iminium trifluoroacetate is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116405-43-7
Molecular Formula C38H30F3NO2P2
Molecular Weight (g/mol) 651.61
MDL Number MFCD15146355
InChI Key NDLHFNFVQBXEPE-UHFFFAOYSA-M
Synonym 1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium 2,2,2-trifluoroacetate, ppn tfa, ksc924o3n, bis triphenylphosphine iminiumtrifluoroacetate, bis-triphenylphosphoranylidene ammonium tfa, bis triphenylphosphine iminium trifluoroacetic acid salt, bis-triphenylphosphoranylidene ammonium tfa ppn tfa, bis triphenyl-??-phosphanylidene azanium trifluoroacetate, 1,1,1-triphenyl-n-triphenylphosphoranylidene-phosphoraniminium 2,2,2-trifluoroacetate, triphenyl-n-triphenyl-lambda∼5∼-phosphanylidene-lambda∼5∼-phosphaniminium trifluoroacetate
PubChem CID 19800386
IUPAC Name 2,2,2-trifluoroacetate;triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium
SMILES [O-]C(=O)C(F)(F)F.C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 782.65
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Bis(triphenylphosphine)iminium trifluoroacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.