missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylstannyl)acetylene, 95%, ACROS Organics™

Product Code. 10506353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10506353 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10506353 Supplier Thermo Scientific Acros Supplier No. 380040050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2117-50-2
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C8H18Sn2
Linear Formula (CH3)3SnCidentical toCSn(CH3)3
MDL Number MFCD01073798
Quantity 5 g
Synonym bis trimethylstannyl acetylene, ethynylenebis trimethylstannane, bis trimethyltin acetylene, 1,2-bis trimethylstannyl ethyne, trimethyl 2-trimethylstannylethynyl stannane, ethyne-1,2-diylbis trimethylstannane, acmc-20allr, ethynylenebis trimethyl tin, bis trimethyltin iv acetylene
InChI Key CDIFRACRLLNHOO-UHFFFAOYSA-N
SMILES C[Sn](C)(C)C#C[Sn](C)(C)C
IUPAC Name trimethyl[2-(trimethylstannyl)ethynyl]stannane
Molecular Weight (g/mol) 351.65
PubChem CID 75031
Formula Weight 351.65
Percent Purity ≥94.0%
Physical Form Crystalline Powder
Chemical Name or Material Bis(trimethylstannyl)acetylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.