missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)sulfide, 95%

Product Code. 10354352 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10354352 5 g Glass bottle 5g
11925851 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10354352 Supplier Thermo Scientific Supplier No. 367370050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3385-94-2 | C6H18SSi2 | 178.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3385-94-2
Molecular Formula C6H18SSi2
Molecular Weight (g/mol) 178.45
MDL Number MFCD00014851
InChI Key RLECCBFNWDXKPK-UHFFFAOYSA-N
Synonym bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8
PubChem CID 76920
IUPAC Name trimethyl(trimethylsilylsulfanyl)silane
SMILES C[Si](C)(C)S[Si](C)(C)C

Specifications

Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 163.0°C (170.0 mmHg)
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula S(Si(CH3)3)2
Refractive Index 1.4555 to 1.4585
Quantity 5 g
Specific Gravity 0.95
Solubility Information Solubility in water: .. Other solubilities: soluble in toluene,ethers,hydrocarbons
Formula Weight 178.45
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl)sulfide, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(trimethylsilyl)sulfide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Bis(trimethylsilyl)sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.