missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl) malonate, 98%

Product Code. 10006293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10006293 50 g Glass bottle 50g
10299793 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10006293 Supplier Thermo Scientific Acros Supplier No. 313300500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18457-04-0 | C9H20O4Si2 | 248.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18457-04-0
Molecular Formula C9H20O4Si2
Molecular Weight (g/mol) 248.43
MDL Number MFCD00009631
InChI Key ATCKJLDGNXGLAO-UHFFFAOYSA-N
Synonym bis trimethylsilyl malonate, malonic acid bis trimethylsilyl ester, propanedioic acid, bis trimethylsilyl ester, malonic acid, bis trimethylsilyl ester, malonic acid ditms, malonic acid 2tms, malonic acid, bis-tms, bis trimethylsilyl malonat, malonic acid bis-tms, acmc-1c3vy
PubChem CID 87656
IUPAC Name bis(trimethylsilyl) propanedioate
SMILES C[Si](C)(C)OC(=O)CC(=O)O[Si](C)(C)C

Specifications

Color Colorless to Straw
Density 0.9720g/mL
Boiling Point 63°C to 66°C (1.0 mmHg)
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiOC(O)CH2C(O)OSi(CH3)3
Refractive Index 1.4150 to 1.417
Quantity 50 g
Specific Gravity 0.972
Solubility Information Solubility in water: reacts
Formula Weight 248.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl) malonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.