missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)carbodiimide, 97%

Product Code. 11439153
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439153 1 g 1g
11449153 5 g 5g
11459153 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439153 Supplier Thermo Scientific Alfa Aesar Supplier No. A11670.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1000-70-0 | C7H18N2Si2 | 186.405 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000-70-0
Molecular Formula C7H18N2Si2
Molecular Weight (g/mol) 186.405
MDL Number MFCD00051538
InChI Key KSVMTHKYDGMXFJ-UHFFFAOYSA-N
Synonym bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
PubChem CID 70473
IUPAC Name N,N'-bis(trimethylsilyl)methanediimine
SMILES C[Si](C)(C)N=C=N[Si](C)(C)C

Specifications

Melting Point -23°C
Density 0.823
Boiling Point 163°C to 164°C
Flash Point 42°C (107°F)
Odor Mild
Linear Formula (CH3)3SiN=C=NSi(CH3)3
Refractive Index 1.434
Quantity 1 g
UN Number UN1993
Beilstein 2042082
Sensitivity Moisture sensitive
Formula Weight 186.41
Percent Purity 97%
Chemical Name or Material Bis (trimethylsilyl)carbodiimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.