missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsiloxy)methylsilane, 97%

Product Code. 15467068
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467068 250 mL 250mL
15457068 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467068 Supplier Thermo Scientific Alfa Aesar Supplier No. H60586.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1873-88-7 | C7H21O2Si3 | 221.498 g/mol

Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

Solubility
Miscible with acetone, ethanol, diethyl ether. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1873-88-7
Molecular Formula C7H21O2Si3
Molecular Weight (g/mol) 221.498
MDL Number MFCD00048000
InChI Key SWGZAKPJNWCPRY-UHFFFAOYSA-N
Synonym 1,1,1,3,5,5,5-heptamethyltrisiloxane, bis trimethylsiloxy methylsilane, unii-rvy9659r8c, trisiloxane, 1,1,1,3,5,5,5-heptamethyl, methyl-bis trimethylsilyloxy silicon, dsstox_cid_18795, methicone 20 cst, unii-6777u11mkt, methylpolysilicone, xiameter mhx-1107 fluid 20 cst
PubChem CID 6327366
IUPAC Name methyl-bis(trimethylsilyloxy)silicon
SMILES C[Si](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Boiling Point 142°C
Quantity 250 mL
Linear Formula [(CH3)3SiO]2SiHCH3
UN Number UN1993
Beilstein 1748455
Flash Point 22°C (72°F)
Solubility Information Miscible with acetone,ethanol,diethyl ether. Insoluble in water.
Formula Weight 222.51
Percent Purity 97%
Refractive Index 1.382
Density 0.819
Chemical Name or Material Bis(trimethylsiloxy)methylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.