missing translation for 'onlineSavingsMsg'
Learn More

[Bis(trifluoroacetoxy)iodo]benzene, 97%

Product Code. 11368166
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368166 5 g 5g
11378166 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368166 Supplier Thermo Scientific Alfa Aesar Supplier No. L15141.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2712-78-9 | C10H5F6IO4 | 430.041 g/mol

[Bis(trifluoroacetoxy)iodo]benzene acts as a reagent in Pummerer-like reactions. It plays an important role in the direct alfa-hydroxylation of ketones under acidic conditions. It is involved in the cyclization of styryl amines to N-alkyl or N-aryl indoles. It serves as a reagent for the chemoselective deprotection of dimethoxybenzyl ethers and tosyloxylation of anilides. Further, it is used in the synthesis of 1,3,4-oxadiazoles by the oxidation of N-acylhydrazones. In addition to this, it is employed in the Hofmann rearrangement for the conversion of cyclobutanecarboxamide to cyclobutylamine hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
[Bis(trifluoroacetoxy)iodo]benzene acts as a reagent in Pummerer-like reactions. It plays an important role in the direct alfa-hydroxylation of ketones under acidic conditions. It is involved in the cyclization of styryl amines to N-alkyl or N-aryl indoles. It serves as a reagent for the chemoselective deprotection of dimethoxybenzyl ethers and tosyloxylation of anilides. Further, it is used in the synthesis of 1,3,4-oxadiazoles by the oxidation of N-acylhydrazones. In addition to this, it is employed in the Hofmann rearrangement for the conversion of cyclobutanecarboxamide to cyclobutylamine hydrochloride.

Solubility
Insoluble in water.

Notes
Light and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2712-78-9
Molecular Formula C10H5F6IO4
Molecular Weight (g/mol) 430.041
MDL Number MFCD00009672
InChI Key PEZNEXFPRSOYPL-UHFFFAOYSA-N
Synonym bis trifluoroacetoxy iodo benzene, pifa, unii-659sfv27xs, bis-trifluoroacetoxyiodobenzene, phenylbis trifluoroacetato-o iodine, bis i,i-trifluoroacetoxy iodobenzene, iodine, phenylbis trifluoroacetato-o, bis trifluoracetoxy-iodobenzene, bis trifluoroacetoxy phenyl iodane
PubChem CID 102317
IUPAC Name [phenyl-(2,2,2-trifluoroacetyl)oxy-$l^{3}-iodanyl] 2,2,2-trifluoroacetate
SMILES C1=CC=C(C=C1)I(OC(=O)C(F)(F)F)OC(=O)C(F)(F)F

Specifications

Melting Point 122°C to 125°C
Quantity 5 g
Beilstein 764767
Sensitivity Light and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 430.04
Percent Purity 97%
Chemical Name or Material [Bis(trifluoroacetoxy)iodo]benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.