missing translation for 'onlineSavingsMsg'
Learn More

Bis(tributyltin) Oxide, TRC

Product Code. 30337053
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30337053 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30337053

Brand: Toronto Research Chemicals TRCB5854305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBTO
CAS 56-35-9
Molecular Formula C24 H54 O Sn2
Quantity 5 g
Synonym TBTO (Bis(tributyltin) oxide), Distannoxane, 1,1,1,3,3,3-hexabutyl- (ACI), 1,1,1,3,3,3-Hexabutyldistannoxane (ACI), Bis(tributyltin) oxide (6CI), Distannoxane, hexabutyl- (8CI, 9CI), 6-Oxa-5,7-distannaundecane, 5,5,7,7-tetrabutyl-, Aceto TBTO, BioMeT 66, BioMeT SRM, BioMeT TBTO, Bis(tributylstannic oxide), Bis(tributylstannyl) ether, Bis(tributylstannyl) oxide, Butinox, Di-tributyl tin oxide, Diall, Hexabutyldistannoxane, Hexabutylditin oxide, Intercide 340A, Lastanox F, Lastanox Q, Lastanox Q1, Lastanox Super, Lastanox T, Lastanox T 20, MT 1E, Mykolastanox F, NSC 22332, NSC 28132, Oxybis(tributylstannane), Oxybis[tributyltin], Stannicide A, Stannicide O, TBTO, TnBTO, Tri-n-butyltin oxide, Tributyltin oxide, Vikol AF 25, Vikol LO 25
InChI Formula InChI=1S/6C4H9.O.2Sn/c6*1-3-4-2;;;/h6*1,3-4H2,2H3;;;
SMILES CCCC[Sn](CCCC)(CCCC)O[Sn](CCCC)(CCCC)CCCC
IUPAC Name tributyl(tributylstannyloxy)stannane
Molecular Weight (g/mol) 596.1
Formula Weight 598.2219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.