missing translation for 'onlineSavingsMsg'
Learn More

Bis(tributylstannyl)acetylene, 97%

Product Code. 10611463 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
5g; Glass bottle
This item is not returnable. View return policy

Product Code. 10611463

Brand: Thermo Scientific Acros 369580050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 994-71-8 | C26H54Sn2 | 604.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 994-71-8
Color Colorless
Density 1.1470g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C26H54Sn2
Linear Formula [CH3(CH2)3]3SnC≡CSn[(CH2)3CH3]3
Refractive Index 1.4920 to 1.4940
MDL Number MFCD00010239
Quantity 5 g
Specific Gravity 1.147
Synonym bis tributylstannyl acetylene, 1,2-bis tributylstannyl ethyne, bis tri-n-butylstannyl acetylene, bis tri-n-butylstannyl ethyne, tributyl 2-tributylstannylethynyl stannane, tributyl tributylstannyl ethynyl stannane #, tributyl 2-tributylstannyl ethynyl stannane
InChI Key CAUONNQGFXOEMY-UHFFFAOYSA-N
SMILES CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC
IUPAC Name tributyl(2-tributylstannylethynyl)stannane
Molecular Weight (g/mol) 604.14
PubChem CID 305981
Formula Weight 604.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Bis(tributylstannyl)acetylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bis(tributylstannyl)acetylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.