missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-o-tolylphosphine)palladium(0), Pd 14.9%

Product Code. 15462457
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462457 1 g 1g
15437095 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462457 Supplier Thermo Scientific Alfa Aesar Supplier No. 045814.03
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

Air Sensitive | CAS: 69861-71-8 | C42H42P2Pd | 715.166 g/mol

Bis(tri-o-tolylphosphine)palladium(0) is used as a catalyst for palladium-catalyzed cross-coupling reactions, Kumada coupling and Heck coupling. It is also used in the amination and arylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69861-71-8
Molecular Formula C42H42P2Pd
Molecular Weight (g/mol) 715.166
MDL Number MFCD12911908
InChI Key CUBIJGNGGJBNOC-UHFFFAOYSA-N
Synonym bis tri-o-tolylphosphine palladium 0, pd o-tol 3p 2, bis tris 2-methylphenyl phosphine palladium, bis tris 2-tolyl phosphine palladium, palladium, bis tris 2-methylphenyl phosphine, bis tris 2-methylphenyl phosphane palladium
PubChem CID 10952654
IUPAC Name palladium;tris(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.[Pd]

Specifications

Melting Point 220°C to 225°C
Color White to Yellow
Assay Percent Range Pd 14.9%
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 715.16
Physical Form Crystalline
Chemical Name or Material Bis(tri-o-tolylphosphine)palladium(0)
compliance-icons
Product Identifier
  • Bis(tri-o-tolylphosphine)palladium(0)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Bis(tri-o-tolylphosphine)palladium(0)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.