Learn More
Bis(tri-o-tolylphosphine)palladium(0), Pd 14.9%
Description
Bis(tri-o-tolylphosphine)palladium(0) is used as a catalyst for palladium-catalyzed cross-coupling reactions, Kumada coupling and Heck coupling. It is also used in the amination and arylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 220°C to 225°C |
| Color | White to Yellow |
| Assay Percent Range | Pd 14.9% |
| Quantity | 250 mg |
| Sensitivity | Air Sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 715.16 |
| Physical Form | Crystalline |
| Chemical Name or Material | Bis(tri-o-tolylphosphine)palladium(0) |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.