missing translation for 'onlineSavingsMsg'
Learn More

Bis[tri(o-tolyl)phosphine]palladium(II) chloride, 95%, ACROS Organics™

Brand:  Acros Organics 377960010

Code : 29

Additional Details : CAS Number : 40691-33-6 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10196993

  • £28.90 / 1g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

Bis[tri(o-tolyl)phosphine]palladium(II) chloride
C42H42Cl2P2Pd
MFCD00274659
95%
786.06

Specifications

Bis[tri(o-tolyl)phosphine]palladium(II) chloride
94% min. (ex Carbon)
Glass bottle
95%
786.06
1g
40691-33-6
Authentic
Crystalline Powder
C42H42Cl2P2Pd
MFCD00274659
Yellow to orange
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bis¬Ętri(o-tolyl)phosphine!palladium(II) chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS