missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-n-butyltin) sulfate, 97%, Thermo Scientific™

Product Code. 11394567
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394567 10 g 10g
11384567 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394567 Supplier Thermo Scientific Alfa Aesar Supplier No. 039624.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26377-04-8
Molecular Formula C24H54O4SSn2
Molecular Weight (g/mol) 676.172
MDL Number MFCD00185908
InChI Key XEPUJTDDAPFGMG-UHFFFAOYSA-L
Synonym bis tri-n-butyltin sulfate, bis tributylstannylium sulfate, hexabutyl, i-sulphato 2--o,o:o',o' ditin
PubChem CID 15787681
IUPAC Name tributylstannanylium;sulfate
SMILES CCCC[Sn+](CCCC)CCCC.CCCC[Sn+](CCCC)CCCC.[O-]S(=O)(=O)[O-]

Specifications

Quantity 10 g
UN Number UN3146
Formula Weight 676.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Bis(tri-n-butyltin) sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.