missing translation for 'onlineSavingsMsg'
Learn More

Bis(tetramethylene)fluoroformamidinium hexafluorophosphate

Product Code. 11384875
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384875 1 g 1g
11394875 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384875 Supplier Thermo Scientific Alfa Aesar Supplier No. L18013.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 164298-25-3 | C9H16F7N2P | 316.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164298-25-3
Molecular Formula C9H16F7N2P
Molecular Weight (g/mol) 316.204
MDL Number MFCD02683430
InChI Key MNJUGQKOHJQOCK-UHFFFAOYSA-N
Synonym btffh, bis tetramethylene fluoroformamidinium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, c9h16fn2.f6p, fluoro-dipyrrolidinocarbenium hexafluorophosphate, 1-fluoro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate t, 1e-1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n inverted exclamation marka,n inverted exclamation marka-bis tetramethylene formamidinium hexafluorophosphate
PubChem CID 10935980
IUPAC Name 1-[fluoro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
SMILES C1CCN(C1)C(=[N+]2CCCC2)F.F[P-](F)(F)(F)(F)F

Specifications

Melting Point ∼155°C
Quantity 1 g
UN Number UN1759
Sensitivity Moisture sensitive
Formula Weight 316.2
Chemical Name or Material Bis(tetramethylene)fluoroformamidinium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.