missing translation for 'onlineSavingsMsg'
Learn More

Bis(tetramethylcyclopentadienyl)nickel(II), 98+%

Product Code. 11382969
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11382969

Brand: Thermo Scientific Alfa Aesar H27784.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79019-60-6 | C18H26Ni | 301.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79019-60-6
Molecular Formula C18H26Ni
Molecular Weight (g/mol) 301.099
MDL Number MFCD01862460
InChI Key PPBIVTWBQQUEKM-UHFFFAOYSA-N
Synonym nickel 2+ bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide, nickel 2+ ion bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide
PubChem CID 73994274
IUPAC Name nickel(2+);1,2,3,5-tetramethylcyclopenta-1,3-diene
SMILES CC1[C-]=C(C(=C1C)C)C.CC1[C-]=C(C(=C1C)C)C.[Ni+2]

Specifications

Melting Point 107°C to 112°C
Quantity 1 g
Sensitivity Air and moisture sensitive
Formula Weight 301.1
Percent Purity ≥98%
Chemical Name or Material Bis(tetramethylcyclopentadienyl)nickel(II)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.