missing translation for 'onlineSavingsMsg'
Learn More

Bis(tert-butylcyclopentadienyl)dimethylhafnium(IV), 98+%, Thermo Scientific Chemicals

Product Code. 11312989
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312989 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11312989 Supplier Thermo Scientific Alfa Aesar Supplier No. H27791.06
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 68193-45-3 | Molecular Formula: C20H32Hf

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68193-45-3
Molecular Formula C20H32Hf
MDL Number MFCD01073804
Synonym 1,1'-Di-tert-butylhafnocene dimethyl; Dimethylbis(tert-butylcyclopentadienyl)hafnium(IV)

Specifications

Melting Point 73°C to 76°C
Quantity 5 g
UN Number UN1325
Formula Weight 450.97
Percent Purity ≥98%
Chemical Name or Material Bis(tert-butylcyclopentadienyl)dimethylhafnium(IV)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.