missing translation for 'onlineSavingsMsg'
Learn More

Bis(pyridine)iodonium tetrafluoroborate, 97%

Product Code. 11368666
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11368666

Brand: Thermo Scientific Alfa Aesar L19385.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15656-28-7 | C10H10BF4IN2 | 371.91 g/mol

Bis(pyridine)iodonium tetrafluoroborate is used as mild iodinating and oxidizing reagent. It is also useful for selective iodination of free phenolic groups in tyrosine residues during solid-phase synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(pyridine)iodonium tetrafluoroborate is used as mild iodinating and oxidizing reagent. It is also useful for selective iodination of free phenolic groups in tyrosine residues during solid-phase synthesis.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15656-28-7
Molecular Formula C10H10BF4IN2
Molecular Weight (g/mol) 371.91
MDL Number MFCD03703393
InChI Key BMDSRCBKJZCUBH-UHFFFAOYSA-N
Synonym bis pyridine iodonium tetrafluoroborate, barluenga's reagent, bis pyridine iodonium i tetrafluoroborate, iodanium pyridine tetrafluoroborate, iodonium pyridine tetrafluoroborate, bispyridine iodonium tetrafluoroborate, bis pyridin iodonium tetrafluoroborate, iodium bis pyridine tetrafluoroborate, bis pyridine-iodonium tetrafluoroborate
PubChem CID 10883201
SMILES F[B-](F)(F)F.[I-]([N+]1=CC=CC=C1)[N+]1=CC=CC=C1

Specifications

Melting Point ∽160°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 4284739
Sensitivity Light and moisture sensitive
Solubility Information Hydrolyzes in water.
IUPAC Name bis(pyridin-1-ium-1-yl)iodanuide; tetrafluoroboranuide
Formula Weight 371.91
Percent Purity 97%
Chemical Name or Material Bis(pyridine)iodonium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.