missing translation for 'onlineSavingsMsg'
Learn More

Bis(phenylthio)methane, 98+%

Product Code. 11391387
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391387 25 g 25g
11381387 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391387 Supplier Thermo Scientific Alfa Aesar Supplier No. L01723.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3561-67-9 | C13H12S2 | 232.359 g/mol

Acyl anion equivalent with some advantages over 1,3-dithiane for the preparation of vinyl sulfides. Bis(phenylthio)methane is used in the synthesis of the dihydroxy thioethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acyl anion equivalent with some advantages over 1,3-dithiane for the preparation of vinyl sulfides. Bis(phenylthio)methane is used in the synthesis of the dihydroxy thioethers.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3561-67-9
Molecular Formula C13H12S2
Molecular Weight (g/mol) 232.359
MDL Number MFCD00003067
InChI Key ZHUPZVIALZHGGP-UHFFFAOYSA-N
Synonym bis phenylthio methane, methylenebis thio bisbenzene, bis phenyithio methane, formaldehyde diphenyl mercaptal, 1,1'-methylenebis thio bisbenzene, phenylsulfanyl methyl sulfanyl benzene, 1-phenylthio methyl thio benzene, bis thiophenoxy methane
PubChem CID 77097
IUPAC Name phenylsulfanylmethylsulfanylbenzene
SMILES C1=CC=C(C=C1)SCSC2=CC=CC=C2

Specifications

Melting Point 36°C to 38°C
Boiling Point 194°C (8 mmHg)
Linear Formula C6H5SCH2SC6H5
Quantity 25 g
Beilstein 1874146
Solubility Information Insoluble in water.
Formula Weight 232.36
Percent Purity ≥98%
Chemical Name or Material Bis(phenylthio)methane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.