missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)titanium dichloride, Thermo Scientific Chemicals

Product Code. 11364467
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11364467

Brand: Thermo Scientific Alfa Aesar 039570.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is used as an intermediate in the synthesis of troticenyl monophosphanes and [2]troticenophanes with C-P and C-Si bridges.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11136-36-0
Molecular Formula C20H30Cl2Ti
Molecular Weight (g/mol) 389.227
MDL Number MFCD00049153
InChI Key ZBFBXTFQCKIUHU-UHFFFAOYSA-L
Synonym titanium 4+ chloride 2,3,4,5,5-pentamethylcyclopenta-1,3-dien-1-ide 1/2/2
PubChem CID 91997586
IUPAC Name 1,2,3,5,5-pentamethylcyclopenta-1,3-diene;titanium(4+);dichloride
SMILES CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Cl-].[Cl-].[Ti+4]

Specifications

Color Red to Brown
Quantity 1 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 389.23
Physical Form Crystalline Solid
Chemical Name or Material Bis(pentamethylcyclopentadienyl)titanium dichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.