missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)nickel

Product Code. 11373628
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373628 1 g 1g
11383628 250 mg 250mg
11393628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373628 Supplier Thermo Scientific Alfa Aesar Supplier No. 039238.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 74507-63-4 | C20H30Ni | 329.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74507-63-4
Molecular Formula C20H30Ni
Molecular Weight (g/mol) 329.153
MDL Number MFCD00058704
InChI Key CHPLEWYRKUFKQP-UHFFFAOYSA-N
Synonym Decamethylnickelocene
PubChem CID 24943158
IUPAC Name nickel(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene
SMILES CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ni+2]

Specifications

Packaging Ampule
Quantity 1 g
Sensitivity Air and moisture sensitive
Formula Weight 329.17
Physical Form Crystalline
Chemical Name or Material Bis(pentamethylcyclopentadienyl)nickel

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.