missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)iron

Product Code. 10347331 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10347331

Brand: Thermo Scientific Acros 347740050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Bis(pentamethylcyclopentadienyl)iron, C20H30Fe, CAS Number-12126-50-0, Decamethylferrocene, 5g, Yellow to Orange, 326.3, MFCD00058736, Authentic, Glass bottle, Crystalline Powder | CAS: 12126-50-0 | C20H30Fe | 326.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12126-50-0
Molecular Formula C20H30Fe
Molecular Weight (g/mol) 326.31
MDL Number MFCD00058736
InChI Key VKRKSGODPDKGAD-UHFFFAOYSA-N
Synonym Decamethylferrocene
SMILES Cc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Fe]2345

Specifications

Melting Point 277°C, 530.6°F
Color Yellow to Orange
Packaging Glass bottle
Quantity 5 g
Formula Weight 326.3
Physical Form Crystalline Powder
Chemical Name or Material Bis(pentamethylcyclopentadienyl)iron

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.