missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct, Thermo Scientific™

Product Code. 11375857
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375857 250 mg 250mg
11355857 1 g 1g
11365857 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375857 Supplier Thermo Scientific Alfa Aesar Supplier No. 043639.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312739-90-5
Molecular Formula C20H30Ba
Molecular Weight (g/mol) 407.787
MDL Number MFCD02684523
InChI Key ZVWNPBSWYPWMIF-UHFFFAOYSA-N
Synonym acmc-1aijw, barium,bis 1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
PubChem CID 21946354
IUPAC Name barium(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene
SMILES CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ba+2]

Specifications

Packaging Ampule
Quantity 250 mg
UN Number UN1564
Formula Weight 479.92
Physical Form Powder
Chemical Name or Material Bis (pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.