missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentafluorophenyl)phenylphosphine, 90%, tech., Thermo Scientific™

Product Code. 10145420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10145420 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10145420 Supplier Thermo Scientific Acros Supplier No. 313171000
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 5074-71-5
Melting Point 56.0°C to 62.0°C
Boiling Point 105.0°C (0.2 mmHg)
Flash Point >110°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C18H5F10P
Linear Formula (C6F5)2PC6H5
Quantity 100 mg
Synonym bis pentafluorophenyl phenylphosphine, dftpp, decafluorotriphenylphosphine, bis perfluorophenyl phenyl phosphine, bis 2,3,4,5,6-pentafluorophenyl-phenylphosphane, phosphine,bis 2,3,4,5,6-pentafluorophenyl phenyl, bis 2,3,4,5,6-pentafluorophenyl phenyl phosphane, ultramark 443, bis pentafluorophenyl phenyl phosphite, bis pentafluorophenyl phenyl phosphine
Solubility Information Solubility in water: insoluble in water.
InChI Key OYNXPGGNQMSMTR-UHFFFAOYSA-N
SMILES FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)-phenylphosphane
Molecular Weight (g/mol) 442.20
PubChem CID 78757
Formula Weight 442.2
Percent Purity 90%
Chemical Name or Material Bis(pentafluorophenyl)phenylphosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.