missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Bis-PEG4-acid

Product Code. 16455676
Encompass_Preferred
Change view
Click to view available options
:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16455676 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16455676 Supplier MedChemExpress Supplier No. HY119429

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bis-PEG4-acid is a PEG PROTAC linker.

Specifications

Molecular Weight (g/mol) 294.3 g/mol
CAS 31127-85-2
Content And Storage Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Quantity 100 mg
Solubility DMSO : 300 mg/mL (1019.37 mM; Need ultrasonic) ∣ Ethanol : 100 mg/mL (339.79 mM; Need ultrasonic)
Target Factor Xa ∣ Thrombin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.