missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(N,N-dibutylamine)-N,N’-disulfide-d36, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Bis(N,N-dibutylamine)-N,N'-disulfide-d36 |
| CAS | 1216478-97-5 |
| Molecular Formula | C16D36N2S2 |
| Quantity | 25 mg |
| Synonym | N,N’-Dithiobis[N-butyl]-1-butanamine-d36, N,N’-Dithiobis-dibutylamine-d36 |
| InChI Formula | InChI=1S/C16H36N2S2/c1-5-9-13-17(14-10-6-2)19-20-18(15-11-7-3)16-12-8-4/h5-16H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])N(SSN(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H] |
| IUPAC Name | N-[[bis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)amino]disulfanyl]-1,1,2,2,3,3,4,4,4-nonadeuterio-N-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)butan-1-amine |
| Molecular Weight (g/mol) | 356.82 |
| Formula Weight | 356.458 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction