missing translation for 'onlineSavingsMsg'
Learn More

Bis-N-Cbz-Lisdexamphetamine, TRC

Product Code. 30385734
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385734 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385734 Supplier Toronto Research Chemicals Supplier No. TRCD41825250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzyl ((R)-6-oxo-6-(((S)-1-phenylpropan-2-yl)amino)hexane-1,5-diyl)dicarbamate
CAS 5002-57-3
Molecular Formula C31 H37 N3 O5
Quantity 50 mg
Synonym Dibenzyl ((R)-6-oxo-6-(((S)-1-phenylpropan-2-yl)amino)hexane-1,5-diyl)dicarbamate, Bis-N-Cbz-Lisdexamphetamine, Z-Lys(Z)-Dextroamphetamine
InChI Formula InChI=1S/C31H37N3O5/c1-24(21-25-13-5-2-6-14-25)33-29(35)28(34-31(37)39-23-27-17-9-4-10-18-27)19-11-12-20-32-30(36)38-22-26-15-7-3-8-16-26/h2-10,13-18,24,28H,11-12,19-23H2,1H3,(H,32,36)(H,33,35)(H,34,37)/t24-,28+/m0/s1
SMILES C[C@@H](Cc1ccccc1)NC(=O)[C@@H](CCCCNC(=O)OCc2ccccc2)NC(=O)OCc3ccccc3
IUPAC Name benzyl N-[(2R)-1-oxo-6-(phenylmethoxycarbonylamino)-1-[[(2S)-1-phenylpropan-2-yl]amino]hexan-2-yl]carbamate
Molecular Weight (g/mol) 531.643
Formula Weight 531.273
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.