missing translation for 'onlineSavingsMsg'
Learn More

Bis(heptafluoroisopropyl)ketone, 97%

Product Code. 11349345
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11349345

Brand: Thermo Scientific Alfa Aesar L16708.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 813-44-5 | C7F14O | 366.054 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-44-5
Molecular Formula C7F14O
Molecular Weight (g/mol) 366.054
MDL Number MFCD00042087
InChI Key GRVMOMUDALILLH-UHFFFAOYSA-N
Synonym bis perfluoroisopropyl ketone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl pentan-3-one, bis perfluorisopropyl ketone, bis heptafluoroisopropyl ketone, 3-pentanone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl, perfluoroisopropyl ketone, bis heptafluoroisopropyl methanone, cf3 2cfc o cf cf3 2, 3-hexanone, 1,1,1,2,4,4,5,5,6,6,6-undecafluoro-2-trifluoromethyl, tert-butyl 2-oxo-1-pyrrolidinecarboxylat
PubChem CID 69941
IUPAC Name 1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one
SMILES C(=O)(C(C(F)(F)F)(C(F)(F)F)F)C(C(F)(F)F)(C(F)(F)F)F

Specifications

Density 1.661
Boiling Point 72°C to 73°C
Quantity 5 g
UN Number UN2810
Beilstein 1916532
Formula Weight 366.06
Percent Purity 97%
Chemical Name or Material Bis(heptafluoroisopropyl)ketone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.