missing translation for 'onlineSavingsMsg'
Learn More

Bis(ethylene)(2,4-pentanedionato)rhodium(I), Rh 39.9% min

Product Code. 11313698
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313698 1 g 1g
11303698 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313698 Supplier Thermo Scientific Alfa Aesar Supplier No. 039288.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12082-47-2 | C9H15O2Rh | 258.12 g/mol

It acts as a hydrogenation precursor. It is also used as intermediates which acts as catalyst such as in olefin hydroformylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12082-47-2
Molecular Formula C9H15O2Rh
Molecular Weight (g/mol) 258.12
MDL Number MFCD00015354
InChI Key FLRBEQQDEGBCJS-FGSKAQBVSA-M
Synonym acetylacetonatobis ethylene rhodium i, bis ethylene 2,4-pentanedionato rhodium i, acetylacetonatobis ethylene rhodium, ethylene; z-4-oxopent-2-en-2-olate; rhodium
PubChem CID 11630270
IUPAC Name ethene;(Z)-4-oxopent-2-en-2-olate;rhodium
SMILES [Rh+].C=C.C=C.C\C([O-])=C\C(C)=O

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in dichloromethane, chloroform
Formula Weight 258.13
Concentration or Composition (by Analyte or Components) Rh 39.9% min.
Physical Form Crystalline
Chemical Name or Material Bis(ethylene)(2,4-pentanedionato)rhodium(I)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.