missing translation for 'onlineSavingsMsg'
Learn More

Bis(diphenylphosphino)methane, 97%

Product Code. 11344579
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344579 2 g 2g
11484577 10 g 10g
11494577 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344579 Supplier Thermo Scientific Alfa Aesar Supplier No. B21112.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2071-20-7 | C25H22P2 | 384.40 g/mol

It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2071-20-7
Molecular Formula C25H22P2
Molecular Weight (g/mol) 384.40
MDL Number MFCD00003537
InChI Key XGCDBGRZEKYHNV-UHFFFAOYSA-N
Synonym bis diphenylphosphino methane, dppm, methylenebis diphenylphosphine, phosphine, methylenebis diphenyl, tetraphenylmethylenediphosphine, unii-i5o2hzr38v, methanediylbis diphenylphosphine, diphenylphosphanyl methyl diphenylphosphane, 1,1-bis diphenylphosphino methane
PubChem CID 74952
IUPAC Name diphenylphosphanylmethyl(diphenyl)phosphane
SMILES C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 118°C to 119°C
Quantity 2 g
Beilstein 758760
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 384.4
Percent Purity 97%
Chemical Name or Material Bis(diphenylphosphino)methane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.