missing translation for 'onlineSavingsMsg'
Learn More

Bis(dimethylamino)phosphoryl Chloride, TRC

Product Code. 30329118
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329118 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329118 Supplier Toronto Research Chemicals Supplier No. TRCB42975510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(dimethylamino)phosphoryl Chloride
CAS 1605-65-8
Molecular Formula C23 H21 N O2
Quantity 10 g
Synonym Bis(dimethylamino)chlorophosphine Oxide, Bis(dimethylamino)phosphinic Chloride, Bis(dimethylamino)phosphinyl Chloride, Bis(dimethylamino)phosphoryl Chloride, Chlorobis(dimethylamino)phosphine Oxide, N,N,N',N'-Tetramethyldiamidophosphorochloridate, NSC 30697, Tetramethyldiamidophosphoryl Chloride, Tetramethylphosphorochloridic Diamide, Tetramethylphosphorodiamidic Chloride, Tetramethylphosphorodiamidoyl Chloride
InChI Formula InChI=1S/C23H21NO2/c1-26-22(25)21-17-24(21)23(18-11-5-2-6-12-18,19-13-7-3-8-14-19)20-15-9-4-10-16-20/h2-16,21H,17H2,1H3
SMILES COC(=O)C1CN1C(c2ccccc2)(c3ccccc3)c4ccccc4
IUPAC Name methyl 1-tritylaziridine-2-carboxylate
Molecular Weight (g/mol) 343.418
Formula Weight 343.157
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.