missing translation for 'onlineSavingsMsg'
Learn More

Bis(dimethylamino)phosphorochloridate, 94%, Thermo Scientific Chemicals

Product Code. 11383678
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383678 2 g 2g
11373678 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383678 Supplier Thermo Scientific Alfa Aesar Supplier No. 039274.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1605-65-8
Molecular Formula C4H12ClN2OP
Molecular Weight (g/mol) 170.58
MDL Number MFCD00008302
InChI Key WYLQARGYFXBZMD-UHFFFAOYSA-N
Synonym bis dimethylamino phosphoryl chloride, phosphorodiamidic chloride, tetramethyl, bis dimethylamino phosphinic chloride, n,n,n',n'-tetramethylphosphorodiamidic chloride, bis dimethylamino phosphorylchloride, bis n,n-dimethylamino phosphinic chloride, ch3 2n 2pocl, bis dimethylamino phosphorochloridate, tetramethylphosphorodiamidic chloride, phosphorodiamidicchloride, n,n,n',n'-tetramethyl
PubChem CID 74150
IUPAC Name N-[chloro(dimethylamino)phosphoryl]-N-methylmethanamine
SMILES CN(C)P(Cl)(=O)N(C)C

Specifications

Color Colorless
Density 1.152 g/mL
Boiling Point 246°C to 247°C
Flash Point 119°C (246°F)
Odor Pungent
Linear Formula [(CH3)2N]2P(O)Cl
Refractive Index 1.4679
Quantity 2 g
UN Number UN3265
Beilstein 471430
Sensitivity Moisture sensitive
Formula Weight 170.58
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Bis(dimethylamino)phosphorochloridate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.