missing translation for 'onlineSavingsMsg'
Learn More

Bis(dimethylamino)phosphorochloridate, 90%, Tech.

Product Code. 10127793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127793 50 g Glass bottle 50g
10377590 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10127793 Supplier Thermo Scientific Supplier No. 172720500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1605-65-8 | C4H12ClN2OP | 170.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1605-65-8
Molecular Formula C4H12ClN2OP
Molecular Weight (g/mol) 170.58
MDL Number MFCD00008302
InChI Key WYLQARGYFXBZMD-UHFFFAOYSA-N
Synonym bis dimethylamino phosphoryl chloride, phosphorodiamidic chloride, tetramethyl, bis dimethylamino phosphinic chloride, n,n,n',n'-tetramethylphosphorodiamidic chloride, bis dimethylamino phosphorylchloride, bis n,n-dimethylamino phosphinic chloride, ch3 2n 2pocl, bis dimethylamino phosphorochloridate, tetramethylphosphorodiamidic chloride, phosphorodiamidicchloride, n,n,n',n'-tetramethyl
PubChem CID 74150
SMILES CN(C)P(Cl)(=O)N(C)C

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 246.0°C to 247.0°C
Flash Point 119°C
Infrared Spectrum Authentic
Assay Percent Range 88 to 112% (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula [(CH3)2N]2P(O)Cl
Refractive Index 1.4650 to 1.4670
Quantity 50 g
Specific Gravity 1.15
IUPAC Name bis(dimethylamino)phosphinoyl chloride
Formula Weight 170.58
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Bis(dimethylamino)phosphorochloridate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(dimethylamino)phosphorochloridate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.