missing translation for 'onlineSavingsMsg'
Learn More

Bis(dimethylamino)dimethylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11464597
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464597 100 g 100g
11454597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464597 Supplier Thermo Scientific Alfa Aesar Supplier No. B21119.22
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3768-58-9 | Molecular Formula: C6H18N2Si | Molecular Weight: 146.309 g/mol

Bis(dimethylamino)-dimethyl silane has been used to silanize pH-sensitive microelectrodes, used to measure the membrane potential in oocytes, in an experimental procedure aimed towards cloning and characterizing human electrogenic Na+-cotransporter isoform (hhNBC). It is also used for microelectrode measurements, in a study conducted to measure carbon dioxide transport through membranes. It is also used in electronic film deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(dimethylamino)-dimethyl silane has been used to silanize pH-sensitive microelectrodes, used to measure the membrane potential in oocytes, in an experimental procedure aimed towards cloning and characterizing human electrogenic Na+-cotransporter isoform (hhNBC). It is also used for microelectrode measurements, in a study conducted to measure carbon dioxide transport through membranes. It is also used in electronic film deposition.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from air and oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away. Protect from electrostatic charges.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3768-58-9
Molecular Formula C6H18N2Si
Molecular Weight (g/mol) 146.309
MDL Number MFCD00008296
InChI Key QULMGWCCKILBTO-UHFFFAOYSA-N
Synonym bis dimethylamino dimethylsilane, silanediamine, hexamethyl, silanediamine, n,n,n',n',1,1-hexamethyl, acmc-1ctl2, bis-dimethylamino dimethylsilane, n,n,n,n-hexamethylsilanediamine #, dimethylsilane, bis n,n-dimethylamino, n,n,n',n',1,1-hexamethylsilanediamine
PubChem CID 77384
IUPAC Name N-[dimethylamino(dimethyl)silyl]-N-methylmethanamine
SMILES CN(C)[Si](C)(C)N(C)C

Specifications

Boiling Point 128°C to 129°C
Melting Point -98°C
Quantity 100 g
Flash Point −7°C (19°F)
Refractive Index 1.417
Sensitivity Moisture sensitive
Density 0.807
UN Number UN2924
Beilstein 1736095
Solubility Information Hydrolyzes in water.
Formula Weight 146.31
Percent Purity 97%
Chemical Name or Material Bis(dimethylamino)dimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.