missing translation for 'onlineSavingsMsg'
Learn More

Bis(dicarbonylcyclopentadienyliron), 99%

Product Code. 11357398
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357398 100 g 100g
11367398 5 g 5g
11377398 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11357398 Supplier Thermo Scientific Alfa Aesar Supplier No. 031114.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 12154-95-9

Cyclopentadienyliron dicarbonyl dimer is a workhorse in organometallic chemistry because it is inexpensive and rugged. It is also used as a pharmaceutical intermediates and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12154-95-9
MDL Number MFCD00135625
Synonym Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl

Specifications

Melting Point 194°C (decomposition)
Color Red
Quantity 100 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Solubility Information It is readily soluble in moderate polar organic solvents such as chloroform and pyridine,but less soluble in carbon tetrachloride and carbon disulfide. It is sparingly soluble in water.
Formula Weight 353.93
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Bis(dicarbonylcyclopentadienyliron)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.