missing translation for 'onlineSavingsMsg'
Learn More

Bis(dibenzylideneacetone)palladium, Thermo Scientific Chemicals

Catalog Number p-68104
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
32005-36-0
C34H28O2Pd
575.02
MFCD00051942
UKSZBOKPHAQOMP-SVLSSHOZSA-N
bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
6505921
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
[Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
32005-36-0
C34H28O2Pd
575.02
MFCD00051942
UKSZBOKPHAQOMP-SVLSSHOZSA-N
bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
6505921
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
[Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150.0°C
Color Black to Brown-Red
Packaging Glass bottle
Linear Formula Pd(C6H5CH=CHC(O)CH=CHC6H5)2
Quantity 1 g
Fieser 15,302
Solubility Information Solubility in water: insoluble.
Formula Weight 575.02
Percent Purity 16 to 21% (Pd)
Physical Form Fine Powder
Chemical Name or Material Bis(dibenzylideneacetone)palladium
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(dibenzylideneacetone)palladium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H290-May be corrosive to metals.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'