missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)zirconium dichloride, 99+%, Thermo Scientific Chemicals

Product Code. 11398368
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398368 5 g 5g
11308378 1 g 1g
11318378 25 g 25g
11388368 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11398368 Supplier Thermo Scientific Alfa Aesar Supplier No. 039558.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconocene dichloride can also be used to prepare the Negishi reagent, Cp2Zr(η2-butene) which is used as a source for Cp2Zr in oxidative cyclisation reactions. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1291-32-3
MDL Number MFCD00003726
Synonym Zirconocene dichloride; Dichlorobis(cyclopentadienyl)zirconium

Specifications

Melting Point 242°C to 245°C
Color White
Boiling Point 124°C to 125°C (15 mmHg)
Quantity 5 g
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 292.32
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Bis(cyclopentadienyl)zirconium dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.