missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)vanadium dichloride, 95%, Thermo Scientific™

Catalog Number 11314507
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
12083-48-6
MFCD00014260
Vanadocene dichloride
This item is not returnable. View return policy
12083-48-6
MFCD00014260
Vanadocene dichloride

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Melting Point 250°C (decomposition)
Color Green
Density 1.60 g/mL
Odor Pungent
Quantity 1 g
UN Number UN3285
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water
Formula Weight 252.04
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Bis(cyclopentadienyl)vanadium dichloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'