missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)vanadium bromide, 98%, Thermo Scientific™

Catalog Number 11428520
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
64815-29-8
C10H10Br3V-5
420.843
MFCD03701567
QHYHAEAMZXFABY-UHFFFAOYSA-K
132984028
cyclopenta-1,3-diene;vanadium;tribromide
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[V].[Br-].[Br-].[Br-]
This item is not returnable. View return policy
64815-29-8
C10H10Br3V-5
420.843
MFCD03701567
QHYHAEAMZXFABY-UHFFFAOYSA-K
132984028
cyclopenta-1,3-diene;vanadium;tribromide
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[V].[Br-].[Br-].[Br-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 221°C to 222°C
Boiling Point 165°C (0.2 mmHg)(sublimation)
Packaging Ampule
Quantity 1 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Formula Weight 261.04
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bis (cyclopentadienyl)vanadium bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'