missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(cyclopentadienyl)vanadium bromide, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Specifications
Tekniske data
| Melting Point | 221°C to 222°C |
| Boiling Point | 165°C (0.2 mmHg)(sublimation) |
| Packaging | Ampule |
| Quantity | 5 g |
| UN Number | UN3261 |
| Sensitivity | Air and moisture sensitive |
| Formula Weight | 261.04 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Bis (cyclopentadienyl)vanadium bromide |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?