missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(cyclopentadienyl)titanium dichloride, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
This metallocene is a common reagent in organometallic and organic synthesis. Titanocene dichloride is used to prepare titanocene pentasulfide. Used as an anticancer drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Melting Point | ∼285°C (decomposition) |
Color | Red |
Density | 1.60 g/mL |
Odor | Odorless |
Quantity | 50 g |
Sensitivity | Air and moisture sensitive |
Merck Index | 14,9482 |
Solubility Information | Reacts in moist air and in water to form HCl . Slightly hydrolysis in water. |
Formula Weight | 249 |
Percent Purity | ≥99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'