missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)titanium dichloride, 97%

Product Code. 10052690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10052690 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10052690 Supplier Thermo Scientific Acros Supplier No. 215760500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1271-19-8 | C10H10Cl2Ti | 249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-19-8
Molecular Formula C10H10Cl2Ti
Molecular Weight (g/mol) 249
MDL Number MFCD00003723
InChI Key MKNXBRLZBFVUPV-UHFFFAOYSA-L
Synonym Titanocene dichloride
PubChem CID 124040768
IUPAC Name cyclopenta-1,3-diene;titanium(2+);dichloride
SMILES [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 287.0°C to 289.0°C
Color Bordeaux to Red
Density 1.6000g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 50 g
Fieser 06,48
Merck Index 15, 9638
Specific Gravity 1.6
Solubility Information Solubility in water: hydrolyses. Other solubilities: soluble in aromatic hydrocarbons, thf, halogenated, solvents, acetonitrile,, slightly soluble in chloroform, ethanol < 1mg/l,, acetone < 1mg/l
Formula Weight 249
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Bis(cyclopentadienyl)titanium dichloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Bis(cyclopentadienyl)titanium dichloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.