missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)titanium dichloride, 97%

Product Code. 11410333
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410333 250 g 250g
11490323 10 g 10g
11400333 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410333 Supplier Thermo Scientific Alfa Aesar Supplier No. A11456.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1271-19-8 | C10H10Cl2Ti-2 | 248.957 g/mol

One-pot synthesis of α-methylene-γ-butyrolactones from benzaldehydes and bromomethylacrylates. Bis-(cyclopentadienyl)-titanium dichloride alkylaluminum complexls acts as catalysts for the polymerization of ethylene. Polyethylene prepared with these catalysts. Complexes of aluminum chloride and methylaluminum dichloride with bis-(cyclopentadienyl)-titanium dichloride as catalysts for the polymerization of ethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
One-pot synthesis of α-methylene-γ-butyrolactones from benzaldehydes and bromomethylacrylates. Bis-(cyclopentadienyl)-titanium dichloride alkylaluminum complexls acts as catalysts for the polymerization of ethylene. Polyethylene prepared with these catalysts. Complexes of aluminum chloride and methylaluminum dichloride with bis-(cyclopentadienyl)-titanium dichloride as catalysts for the polymerization of ethylene.

Solubility
Slightly soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-19-8
Molecular Formula C10H10Cl2Ti-2
Molecular Weight (g/mol) 248.957
MDL Number MFCD00003723
InChI Key MKNXBRLZBFVUPV-UHFFFAOYSA-L
Synonym Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
PubChem CID 124040768
IUPAC Name cyclopenta-1,3-diene;titanium(2+);dichloride
SMILES [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]

Specifications

Melting Point ∼285°C (decomposition)
Density 1.6
Odor Odorless
Quantity 250 g
Sensitivity Air and moisture sensitive
Merck Index 14,9482
Solubility Information Slightly soluble in water
Formula Weight 249
Percent Purity 97%
Chemical Name or Material Bis (cyclopentadienyl)titanium dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.