missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)ruthenium, Ru 43.2% min, Thermo Scientific Chemicals

£163.00 - £551.00

Chemical Identifiers

CAS 1287-13-4
MDL Number MFCD00001442
Synonym Ruthenocene
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377547
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012781.03
1 g
£163.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11367547
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012781.06
5 g
£551.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

1287-13-4
MFCD00001442
Ruthenocene

Specifications

1287-13-4
MFCD00001442
Ruthenocene
Ru 43.2% min.
Bis(cyclopentadienyl)ruthenium
194°C to 198°C
Air and moisture sensitive
231.26
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 215-065-2

missing translation for 'tsca' : No

missing translation for 'storageNote' :

  • Packaged under argon

Recommended Storage : Ambient temperatures

Product Identifier
  • Bis(cyclopentadienyl)ruthenium

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only