missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)osmium, Thermo Scientific™

Product Code. 11363628
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363628 250 mg 250mg
11353628 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363628 Supplier Thermo Scientific Alfa Aesar Supplier No. 039235.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1273-81-0
Molecular Formula C10H10Os
Molecular Weight (g/mol) 320.42
MDL Number MFCD00058734
InChI Key IEHQTQKQMXEQRM-UHFFFAOYSA-N
Synonym osmocene
PubChem CID 22391851
IUPAC Name cyclopenta-1,3-diene;osmium(2+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Os+2]

Specifications

Melting Point 226°C to 228°C
Quantity 250 mg
Sensitivity Air and moisture sensitive
Formula Weight 320.39
Physical Form Crystalline
Chemical Name or Material Bis(cyclopentadienyl)osmium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.