missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)nickel, dry

Product Code. p-7080387
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 15442977

Brand: Thermo Scientific Alfa Aesar 087899.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1271-28-9 | C10H10Ni | 188.883 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-28-9
Molecular Formula C10H10Ni
Molecular Weight (g/mol) 188.883
MDL Number MFCD00001441
InChI Key RQKPFSQDBAZFJV-UHFFFAOYSA-N
Synonym bis cyclopentadienyl nickel ii
PubChem CID 24942185
IUPAC Name cyclopenta-1,3-diene;nickel(2+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Ni+2]

Specifications

Melting Point 171°C to 173°C
Color Green
Quantity 2 g
UN Number UN3391
Sensitivity Air and moisture sensitive
Formula Weight 188.9
Physical Form Crystalline
Chemical Name or Material Bis (cyclopentadienyl)nickel, Dry

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.