missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals

Product Code. 15461067
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15461067 Supplier Thermo Scientific Alfa Aesar Supplier No. 014535.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73138-26-8 | Molecular Formula: C10H10Mn | Molecular Weight: 185.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73138-26-8
Molecular Formula C10H10Mn
Molecular Weight (g/mol) 185.13
MDL Number MFCD00045468
InChI Key LCGVCXIFXLGLHG-UHFFFAOYSA-N
Synonym bis cyclopentadienyl manganese, acmc-20aln3, dicyclopentadienylmanganese, manganese,di-2,4-cyclopentadien-1-yl, bis cyclopenta-1,3-dien-1-yl manganese
PubChem CID 21960473
IUPAC Name cyclopenta-1,3-diene;manganese(2+)
SMILES [Mn](c1cccc1)c1cccc1

Specifications

Melting Point 175°C
Boiling Point 245°C
Flash Point 52°C (126°F)
Packaging Ampule
Quantity 5 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Formula Weight 185.13
Physical Form Powder
Chemical Name or Material Bis(cyclopentadienyl)manganese, Sublimed

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.