missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)dimethyltitanium, 5 wt.% in toluene, AcroSeal™

Product Code. 10591293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10591293 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10591293 Supplier Thermo Scientific Acros Supplier No. 301761000
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1271-66-5 | C12H20Ti | 212.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Specifications

CAS 1271-66-5, 108-88-3
Color Orange to Red
Density 0.9g/mL
Packaging AcroSeal™ Glass Bottle
Molecular Formula C12H20Ti
MDL Number MFCD02093882
Quantity 100 mL
Synonym bis cyclopentadienyl dimethyltitanium, dimethyltitanocene in tetrahydrofuran/toluene
InChI Key CRKLPNOLYBORCP-UHFFFAOYSA-N
SMILES C.C.C1C=CC=C1.C1C=CC=C1.[Ti]
IUPAC Name cyclopenta-1,3-diene;methane;titanium
Molecular Weight (g/mol) 212.159
PubChem CID 131674729
Formula Weight 208.14
Physical Form Solution
Chemical Name or Material Bis(cyclopentadienyl)dimethyltitanium, 5 wt. % solution in Toluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bis(cyclopentadienyl)dimethyltitanium, 5 wt% solution in toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.