missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)chromium, sublimed, 97+%

Product Code. 11346199
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346199 5 g 5g
11336199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346199 Supplier Thermo Scientific Alfa Aesar Supplier No. 043177.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1271-24-5 | C10H10Cr | 182.19 g/mol

Provides catalyst for ethylene polymerization. A precursor to other chromium(II) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-24-5
Molecular Formula C10H10Cr
Molecular Weight (g/mol) 182.19
MDL Number MFCD00013750
InChI Key TYYBBNOTQFVVKN-UHFFFAOYSA-N
Synonym Chromocene
IUPAC Name Bis(cyclopentadienyl)chromium
SMILES [Cr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1

Specifications

Melting Point 172°C to 173°C
Color Red
Quantity 5 g
UN Number UN3391
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 182.18
Percent Purity ≥97%
Physical Form Crystalline
Chemical Name or Material Bis (cyclopentadienyl)chromium, Sublimed

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.